4,5-Dibromo-1,2-phenylenediamine 98,0

Supplier: TCI America

Synonyms: 1,2-Diamino-4,5-dibromobenzene, 4,5-Dibromobenzene-1,2-diamine

D4062-5G D4062-25G
TCD4062-5GEA 201.61 CAD
TCD4062-5G TCD4062-25G
4,5-Dibromo-1,2-phenylenediamine 98,0
4,5-Dibromo-1,2-phenylenediamine

CAS Number: 49764-63-8
Molecular Formula: C6H6Br2N2
Molecular Weight: 265.94
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 137


Other Product Names / Synonyms: 1,2-Diamino-4,5-dibromobenzene

Formula: C₆H₆Br₂N₂
MW: 265.94 g/mol
Melting Pt: 137 °C
MDL Number: MFCD00795598
CAS Number: 49764-63-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce2df072949c28fb4454be308e54842c24b20cca&vid=ce2df072949c28fb4454be308e54842c24b20cca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR