Diacerein ≥98.0% (by HPLC)
Supplier: TCI America
D4061-1G
D4061-5G
TCD4061-1GEA
199.59
CAD
TCD4061-1G
TCD4061-5G
Diacerein ≥98.0% (by HPLC)
Diacerein
CAS Number: 13739-02-1
MDL Number: MFCD00468030
Molecular Formula: C19H12O8
Molecular Weight: 368.30
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 243
Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Anti-inflammatory Ingredients, Anti-inflammatory Ingredients
Other Product Names / Synonyms: 4,5-Diacetoxyanthraquinone-2-carboxylic Acid
Formula:
C₁₉H₁₂O₈ Melting Pt: 243 °C |
MDL Number:
MFCD00468030 CAS Number: 13739-02-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...