2,5-Dibromo-4-picoline ≥98.0%

Supplier: TCI America

Synonyms: 2,5-Dibromo-4-methylpyridine

D4058-1G D4058-5G
TCD4058-1GEA 44.24 CAD
TCD4058-1G TCD4058-5G
2,5-Dibromo-4-picoline ≥98.0%
2,5-Dibromo-4-picoline

CAS Number: 3430-26-0
MDL Number: MFCD00234955
Molecular Formula: C6H5Br2N
Molecular Weight: 250.92
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 40


Other Product Names / Synonyms: 2,5-Dibromo-4-picoline

Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Melting Pt: 40 °C
MDL Number: MFCD00234955
CAS Number: 3430-26-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac97214b3f8dec170fcd3b9644ba16d333db7c74&vid=ac97214b3f8dec170fcd3b9644ba16d333db7c74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR