Dibenzothiophene-4-boronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 4-Dibenzothiopheneboronic acid

D4057-1G D4057-5G
TCD4057-1GEA 51.19 CAD
TCD4057-1G TCD4057-5G
Dibenzothiophene-4-boronic acid (contains varying amounts of Anhydride)
Dibenzothiophene-4-boronic acid

CAS Number: 108847-20-7
MDL Number: MFCD01318182
Molecular Formula: C12H9BO2S
Molecular Weight: 228.07
Form: Crystal
Color: White
Melting point (°C): 339

Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Melting Pt: 339 °C
MDL Number: MFCD01318182
CAS Number: 108847-20-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84ded430dd7dd7e4bf83b6b99d083cb49cd37e55&vid=84ded430dd7dd7e4bf83b6b99d083cb49cd37e55&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR