5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine, 2,5-Dibromo-3,4-ethylenedioxythiophene
CAS Number: 174508-31-7
MDL Number: MFCD01651770
Molecular Formula: C6H4Br2O2S
Molecular Weight: 299.96
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 96
Storage Temperature: >-20°C
Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 5,7-Dibromo-2,3-dihydrothieno[3,4-b][1,4]dioxine
Formula:
C6H4Br2O2S MW: 299.9700 g/mol Melting Pt: 96 °C |
MDL Number:
MFCD01651770 CAS Number: 174508-31-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...