9,9-Dimethylxanthene ≥98.0% (by GC)

Supplier: TCI America

D4055-5G D4055-1G
TCD4055-5GEA 155.63 CAD
TCD4055-5G TCD4055-1G
9,9-Dimethylxanthene ≥98.0% (by GC)
9,9-Dimethylxanthene

CAS Number: 19814-75-6
MDL Number: MFCD00134434
Molecular Formula: C15H14O
Molecular Weight: 210.28
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 115
Melting point (°C): 38
Flash Point (°C): 113

Formula: C₁₅H₁₄O
Boiling Pt: 115 °C
Melting Pt: 38 °C
Flash Pt: 113 °C
MDL Number: MFCD00134434
CAS Number: 19814-75-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9f88aa96abf76ef12b287d0a2bc1ee491d505178&vid=9f88aa96abf76ef12b287d0a2bc1ee491d505178&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR