2,5-Dibromo-3-octylthiophene ≥98.0% (by GC)
Supplier: TCI America
D4053-5G
TCD4053-5GEA
431.34
CAD
TCD4053-5G
2,5-Dibromo-3-octylthiophene ≥98.0% (by GC)
2,5-Dibromo-3-octylthiophene
CAS Number: 149703-84-4
MDL Number: MFCD00672013
Molecular Formula: C12H18Br2S
Molecular Weight: 354.14
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Flash Point (°C): 110
Specific Gravity (20/20): 1.45
Storage Temperature: 0-10°C
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...