5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0%

Supplier: TCI America

Synonyms: 2,5-Bis(5-bromo-2-thienyl)thiophene

D4050-1G D4050-5G
TCD4050-1GEA 262.28 CAD
TCD4050-1G TCD4050-5G
5,5''-Dibromo-2,2':5',2''-terthiophene ≥97.0%
5,5''-Dibromo-2,2':5',2''-terthiophene

CAS Number: 98057-08-0
MDL Number: MFCD00219109
Molecular Formula: C12H6Br2S3
Molecular Weight: 406.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 158


Product Categories: Materials Chemistry, Building Blocks for Semiconducting Polymers, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 2,5-Bis(5-bromo-2-thienyl)thiophene

Formula: C₁₂H₆Br₂S₃
MW: 406.17 g/mol
Melting Pt: 158 °C
MDL Number: MFCD00219109
CAS Number: 98057-08-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76c7671c5edc8022e2bc47c1aec7cf6c12484b63&vid=76c7671c5edc8022e2bc47c1aec7cf6c12484b63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR