4,5-Dichloro-2-nitroaniline ≥98.0% (by GC)

Supplier: TCI America

D4041-25G D4041-5G
TCD4041-25GEA 218.2 CAD
TCD4041-25G TCD4041-5G
4,5-Dichloro-2-nitroaniline ≥98.0% (by GC)
4,5-Dichloro-2-nitroaniline

CAS Number: 6641-64-1
MDL Number: MFCD00007770
Molecular Formula: C6H4Cl2N2O2
Molecular Weight: 207.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 179

Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 179 °C
MDL Number: MFCD00007770
CAS Number: 6641-64-1
UN: 2237
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23e3c5ccf8abdda7785fabe62899a2ff8321d6fc&vid=23e3c5ccf8abdda7785fabe62899a2ff8321d6fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR