3,4-Diaminofurazan ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,4-Diamino-1,2,5-oxadiazole, 1,2,5-Oxadiazole-3,4-diamine

D4030-1G D4030-5G
TCD4030-1GEA 161.95 CAD
TCD4030-1G TCD4030-5G
3,4-Diaminofurazan ≥98.0% (by GC)
3,4-Diaminofurazan

CAS Number: 17220-38-1
MDL Number: MFCD00138084
Molecular Formula: C2H4N4O
Molecular Weight: 100.08
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 182


Other Product Names / Synonyms: 3,4-Diamino-1,2,5-oxadiazole

Formula: C₂H₄N₄O
MW: 100.08 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00138084
CAS Number: 17220-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=455a15cebeae7180866e93b44637d2d26feb0bee&vid=455a15cebeae7180866e93b44637d2d26feb0bee&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR