1,1-Dimethylpiperidinium chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Mepiquat Chloride, 1,1-Dimethylpiperidin-1-ium chloride
D4016-1G
D4016-5G
TCD4016-1GEA
293.88
CAD
TCD4016-1G
TCD4016-5G
1,1-Dimethylpiperidinium chloride ≥98.0% (by titrimetric analysis)
1,1-Dimethylpiperidinium chloride
CAS Number: 24307-26-4
MDL Number: MFCD00055543
Molecular Formula: C7H16ClN
Molecular Weight: 149.66
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Product Categories: Bioscience, Other Plant Hormones
Other Product Names / Synonyms: Mepiquat Chloride
Formula:
C₇H₁₆ClN MW: 149.66 g/mol |
MDL Number:
MFCD00055543 CAS Number: 24307-26-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...