Daminozide (succinic acid mono(2,2-dimethylhydrazide)) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D4015-25G D4015-5G
TCD4015-25GEA 352.02 CAD
TCD4015-25G TCD4015-5G
Daminozide (succinic acid mono(2,2-dimethylhydrazide)) ≥98.0% (by GC, titration analysis)
Daminozide (Succinic acid mono(2,2-dimethylhydrazide))

CAS Number: 1596-84-5
MDL Number: MFCD00002787
Molecular Formula: C6H12N2O3
Molecular Weight: 160.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 155


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Plant Hormones, Epigenetics Research Reagents
Other Product Names / Synonyms: Succinic Acid 2,2-Dimethylhydrazide

Formula: C₆H₁₂N₂O₃
Melting Pt: 155 °C
MDL Number: MFCD00002787
CAS Number: 1596-84-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4e693d5f6a98a8772e8e2b6b72920d2fc29da3&vid=ba4e693d5f6a98a8772e8e2b6b72920d2fc29da3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR