3-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥98.0% (contains varying amounts of Anhydride)
Supplier: TCI America
D4013-1G
D4013-5G
TCD4013-1GEA
156.42
CAD
TCD4013-1G
TCD4013-5G
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride ≥98.0% (contains varying amounts of Anhydride)
3-(N,N-Dimethylamino)phenylboronic acid hydrochloride
CAS Number: 1256355-23-3
MDL Number: MFCD04112542
Molecular Formula: C8H12BNO2
Molecular Weight: 201.46
Form: Crystal
Color: White
Other Product Names / Synonyms: 3-(Dimethylamino)benzeneboronic Acid Hydrochloride
Formula:
C₈H₁₃BClNO₂ MW: 201.4583 g/mol |
MDL Number:
MFCD04112542 CAS Number: 1256355-23-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...