Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate ≥98.0% (by GC)

Supplier: TCI America

D4012-25G D4012-5G
TCD4012-25GEA 644.64 CAD
TCD4012-25G TCD4012-5G
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate ≥98.0% (by GC)
Diethyl-3,5-dimethyl-2,4-pyrroledicarboxylate

CAS Number: 2436-79-5
MDL Number: MFCD00005218
Molecular Formula: C12H17NO4
Molecular Weight: 239.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 136


Other Product Names / Synonyms: 3,5-Dimethyl-2,4-pyrroledicarboxylic Acid Diethyl Ester

Formula: C₁₂H₁₇NO₄
Melting Pt: 136 °C
MDL Number: MFCD00005218
CAS Number: 2436-79-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03ba295ef74b862a1926d1402c8f5486cb91dddb&vid=03ba295ef74b862a1926d1402c8f5486cb91dddb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR