(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine ≥98.0%

Supplier: TCI America

Synonyms: (S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene, (S)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine, (S)-3-(Dimethylamino)-1-(2-thienyl)-1-propanol

D4010-1G D4010-5G
TCD4010-1GEA 36.34 CAD
TCD4010-1G TCD4010-5G
(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine ≥98.0%
(S)-(-)-N,N-Dimethyl-3-hydroxy-3-(2-thienyl)propanamine

CAS Number: 132335-44-5
MDL Number: MFCD07782107
Molecular Formula: C9H15NOS
Molecular Weight: 185.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79
Specific rotation [a]20/D: -6 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene

Formula: C9H15NOS
MW: 185.2865 g/mol
Melting Pt: 79 °C
MDL Number: MFCD07782107
CAS Number: 132335-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a415b0014aae6f9aabfe9e4d7f68ba92a85a2992&vid=a415b0014aae6f9aabfe9e4d7f68ba92a85a2992&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR