3-(Dimethylamino)-1-(2-thienyl)-1-propanol ≥98.0%

Supplier: TCI America

Synonyms: 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene

D4009-1G D4009-5G
TCD4009-1GEA 99.54 CAD
TCD4009-1G TCD4009-5G
3-(Dimethylamino)-1-(2-thienyl)-1-propanol ≥98.0%
3-(Dimethylamino)-1-(2-thienyl)-1-propanol

CAS Number: 13636-02-7
MDL Number: MFCD09701963
Molecular Formula: C9H15NOS
Molecular Weight: 185.29
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 290
Melting point (°C): 72
Flash Point (°C): 129


Other Product Names / Synonyms: 2-[3-(Dimethylamino)-1-hydroxypropyl]thiophene

Formula: C₉H₁₅NOS
MW: 185.29 g/mol
Boiling Pt: 290 °C
Melting Pt: 72 °C
Flash Pt: 129 °C
MDL Number: MFCD09701963
CAS Number: 13636-02-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=621944b69b74b6de2de0594f8a124f37326beab3&vid=621944b69b74b6de2de0594f8a124f37326beab3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR