2,6-Difluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: 2,6-Difluoropyridine-3-boronic acid

D4006-1G D4006-5G
TCD4006-1GEA 53.72 CAD
TCD4006-1G TCD4006-5G
2,6-Difluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
2,6-Difluoro-3-pyridineboronic acid

CAS Number: 136466-94-9
MDL Number: MFCD03788245
Molecular Formula: C5H4BF2NO2
Molecular Weight: 158.90
Form: Crystal
Color: White

Formula: C₅H₄BF₂NO₂
MW: 158.90 g/mol
MDL Number: MFCD03788245
CAS Number: 136466-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f344a16cd747784921b97f15f90a969980aee47&vid=0f344a16cd747784921b97f15f90a969980aee47&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR