2,4-Dihydroxy-3-nitropyridine ≥98.0% (by GC)

Supplier: TCI America

D4002-25G D4002-5G
TCD4002-25GEA 219.78 CAD
TCD4002-25G TCD4002-5G
2,4-Dihydroxy-3-nitropyridine ≥98.0% (by GC)
2,4-Dihydroxy-3-nitropyridine

CAS Number: 89282-12-2
MDL Number: MFCD01075671
Molecular Formula: C5H4N2O4
Molecular Weight: 156.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 264


Other Product Names / Synonyms: 3-Nitro-2,4-pyridinediol, 4-Hydroxy-3-nitro-2-pyridone, 4-Hydroxy-3-nitro-2(1H)-pyridinone

Formula: C₅H₄N₂O₄
Melting Pt: 264 °C
MDL Number: MFCD01075671
CAS Number: 89282-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b6c09bc08e661bc1b9c26fa815bfa301a6cc30e&vid=3356c9226c4829038e48681e481194d445807ebf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR