Diethyl (4-methoxybenzyl)phosphonate ≥95.0%

Supplier: TCI America

Synonyms: Diethyl 4-methoxybenzylphosphonate

D4000-25G D4000-5G
TCD4000-25GEA 341.28 CAD
TCD4000-25G TCD4000-5G
Diethyl (4-methoxybenzyl)phosphonate ≥95.0%
Diethyl (4-methoxybenzyl)phosphonate

CAS Number: 1145-93-3
MDL Number: MFCD00015662
Molecular Formula: C12H19O4P
Molecular Weight: 258.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 151
Flash Point (°C): 110
Specific Gravity (20/20): 1.14


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Methoxybenzyl)phosphonic Acid Diethyl Ester, 4-(Diethylphosphonomethyl)anisole

Formula: C₁₂H₁₉O₄P
MW: 258.25 g/mol
Boiling Pt: 151 °C
Density: 1.14 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00015662
CAS Number: 1145-93-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=764f9d108ead6cc3326023f0c0e67e5fbd6d357c&vid=764f9d108ead6cc3326023f0c0e67e5fbd6d357c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR