3,5-Dibromo-2-methylthiophene ≥95.0% (by GC)
Supplier: TCI America
D3999-25G
D3999-5G
TCD3999-25GEA
576.86
CAD
TCD3999-25G
TCD3999-5G
3,5-Dibromo-2-methylthiophene ≥95.0% (by GC)
3,5-Dibromo-2-methylthiophene
CAS Number: 29421-73-6
MDL Number: MFCD07781197
Molecular Formula: C5H4Br2S
Molecular Weight: 255.96
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 230
Melting point (°C): -15
Specific Gravity (20/20): 2.00
Storage Temperature: 0-10°C
Formula:
C₅H₄Br₂S Boiling Pt: 230 °C Melting Pt: –15 °C Density: 2.00 (20 °C) |
MDL Number:
MFCD07781197 CAS Number: 29421-73-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...