2,4-Dimethoxy-1,3,5-triazine ≥98.0% (by GC)

Supplier: TCI America

D3998-1G D3998-5G
TCD3998-1GEA 109.02 CAD
TCD3998-1G TCD3998-5G
2,4-Dimethoxy-1,3,5-triazine ≥98.0% (by GC)
2,4-Dimethoxy-1,3,5-triazine

CAS Number: 1898-72-2
Molecular Formula: C5H7N3O2
Molecular Weight: 141.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59

Formula: C₅H₇N₃O₂
Melting Pt: 59 °C
MDL Number: MFCD20257908
CAS Number: 1898-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2baf626de52d406b0ebb9c1da6d94a9b63393c0&vid=e2baf626de52d406b0ebb9c1da6d94a9b63393c0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR