5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D3997-1G D3997-5G
TCD3997-1GEA 67.78 CAD
TCD3997-1G TCD3997-5G
5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride ≥98.0% (by HPLC, total nitrogen)
5,6-Dimethoxy-2-(4-piperidinylmethyl)-1-indanone hydrochloride

CAS Number: 120013-39-0
Molecular Formula: C17H23NO3
Molecular Weight: 325.83
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 250

Formula: C₁₇H₂₄ClNO₃
Melting Pt: 250 °C
MDL Number: MFCD09033179
CAS Number: 120013-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f839f6d51663a32e80214d81673772921da613c3&vid=f839f6d51663a32e80214d81673772921da613c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR