Diethyl-2,5-dibromoterephthalate ≥98.0%
Supplier: TCI America
Synonyms:
2,5-Dibromoterephthalic Acid Diethyl Ester, Diethyl 2,5-dibromoterephthalate
D3995-25G
D3995-5G
TCD3995-25GEA
297.04
CAD
TCD3995-25G
TCD3995-5G
Diethyl-2,5-dibromoterephthalate ≥98.0%
Diethyl-2,5-dibromoterephthalate
CAS Number: 18013-97-3
MDL Number: MFCD12407163
Molecular Formula: C12H12Br2O4
Molecular Weight: 380.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 335
Melting point (°C): 129
Other Product Names / Synonyms: 2,5-Dibromoterephthalic Acid Diethyl Ester
Formula:
C₁₂H₁₂Br₂O₄ Boiling Pt: 335 °C Melting Pt: 129 °C |
MDL Number:
MFCD12407163 CAS Number: 18013-97-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...