2,5-Dibromoterephthalic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3994-25G D3994-5G
TCD3994-25GEA 436.08 CAD
TCD3994-25G TCD3994-5G
2,5-Dibromoterephthalic acid ≥98.0% (by GC, titration analysis)
2,5-Dibromoterephthalic acid

CAS Number: 13731-82-3
MDL Number: MFCD00204593
Molecular Formula: C8H4Br2O4
Molecular Weight: 323.92
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 318

Formula: C₈H₄Br₂O₄
Melting Pt: 318 °C
MDL Number: MFCD00204593
CAS Number: 13731-82-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0778dc5c7dc1e6ede01c9fe50ab9051cdfbdc874&vid=0778dc5c7dc1e6ede01c9fe50ab9051cdfbdc874&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR