6,6'-Dibromo-2,2'-bipyridyl ≥95.0% (by GC)

Supplier: TCI America

D3988-1G D3988-5G
TCD3988-1GEA 226.7 CAD
TCD3988-1G TCD3988-5G
6,6'-Dibromo-2,2'-bipyridyl ≥95.0% (by GC)
6,6'-Dibromo-2,2'-bipyridyl

CAS Number: 49669-22-9
MDL Number: MFCD00234587
Molecular Formula: C10H6Br2N2
Molecular Weight: 313.98
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 225
Lambda max.: 304 nm (EtOH)


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6,6'-Dibromo-2,2'-bipyridine

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR