4,5-Dihydroxyanthraquinone-2-carboxylic acid ≥95.0%

Supplier: TCI America

Synonyms: Rhein, Rhubarb Yellow, Cassic Acid, 4,5-Dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid

D3986-1G D3986-200MG
TCD3986-1GEA 193.39 CAD
TCD3986-1G TCD3986-200MG
4,5-Dihydroxyanthraquinone-2-carboxylic acid ≥95.0%
4,5-Dihydroxyanthraquinone-2-carboxylic acid

CAS Number: 478-43-3
MDL Number: MFCD00009618
Molecular Formula: C15H8O6
Molecular Weight: 284.22
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 321
Lambda max.: 432 nm (MeOH)


Product Categories: Bioscience, Others (Phytochemicals)
Other Product Names / Synonyms: Rhein, Cassic Acid, Rhubarb Yellow

Formula: C₁₅H₈O₆
MW: 284.22 g/mol
Melting Pt: 321 °C
MDL Number: MFCD00009618
CAS Number: 478-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=840f1db8cfb4a2f4408b6cedef6497d7c9ecf9c3&vid=840f1db8cfb4a2f4408b6cedef6497d7c9ecf9c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR