2,6-Difluoro-4-hydroxybenzoic acid ≥98.0%
Supplier: TCI America
D3985-1G
D3985-5G
TCD3985-1GEA
84.37
CAD
TCD3985-1G
TCD3985-5G
2,6-Difluoro-4-hydroxybenzoic acid ≥98.0%
2,6-Difluoro-4-hydroxybenzoic acid
CAS Number: 214917-68-7
MDL Number: MFCD03094500
Molecular Formula: C7H4F2O3
Molecular Weight: 174.10
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 176
Formula:
C₇H₄F₂O₃ Melting Pt: 176 °C |
MDL Number:
MFCD03094500 CAS Number: 214917-68-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...