Dimethyl-2-(1,3-dithiole)phosphonate ≥97.0% (by GC)

Supplier: TCI America

D3981-1G D3981-5G
TCD3981-1GEA 527.72 CAD
TCD3981-1G TCD3981-5G
Dimethyl-2-(1,3-dithiole)phosphonate ≥97.0% (by GC)
Dimethyl-2-(1,3-dithiole)phosphonate

CAS Number: 133113-76-5
Molecular Formula: C5H9O3PS2
Molecular Weight: 212.22
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.42
Storage Temperature: <0°C


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination, Materials Chemistry, Molecular Conductors
Other Product Names / Synonyms: 2-(1,3-Dithiole)phosphonic Acid Dimethyl Ester

Formula: C₅H₉O₃PS₂
Density: 1.42 (20 °C)
CAS Number: 133113-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3786217d9e77ce578b8f44137d85e903e2333266&vid=3786217d9e77ce578b8f44137d85e903e2333266&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR