4-(2,4-Difluorobenzoyl)piperidine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D3979-25G D3979-5G
TCD3979-25GEA 254.38 CAD
TCD3979-25G TCD3979-5G
4-(2,4-Difluorobenzoyl)piperidine hydrochloride ≥98.0% (by HPLC, total nitrogen)
4-(2,4-Difluorobenzoyl)piperidine hydrochloride

CAS Number: 106266-04-0
MDL Number: MFCD01313310
Molecular Formula: C12H13F2NO
Molecular Weight: 261.70
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal

Formula: C₁₂H₁₃F₂NO
MDL Number: MFCD01313310
CAS Number: 106266-04-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c34578a62f9103bccc36cbafb9bfe9e0494b70c5&vid=c34578a62f9103bccc36cbafb9bfe9e0494b70c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR