9,10-Di(1-naphthyl)anthracene ≥98.0%

Supplier: TCI America

Synonyms: 9,10-Di(naphthalen-1-yl)anthracene

D3975-1G D3975-5G
TCD3975-1GEA 137.46 CAD
TCD3975-1G TCD3975-5G
9,10-Di(1-naphthyl)anthracene ≥98.0%
9,10-Di(1-naphthyl)anthracene

CAS Number: 26979-27-1
MDL Number: MFCD04222155
Molecular Formula: C34H22
Molecular Weight: 430.55
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 360

Formula: C₃₄H₂₂
MW: 430.55 g/mol
Melting Pt: 360 °C
MDL Number: MFCD04222155
CAS Number: 26979-27-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd21837516c753b66299f241dd3c61aea4fda18f&vid=dd21837516c753b66299f241dd3c61aea4fda18f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR