9,9-Dimethylfluoren-2-boronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(9,9-Dimethyl-9H-fluoren-2-yl)boronic acid
D3974-1G
D3974-5G
TCD3974-1GEA
72.68
CAD
TCD3974-1G
TCD3974-5G
9,9-Dimethylfluoren-2-boronic acid (contains varying amounts of Anhydride)
9,9-Dimethylfluoren-2-boronic acid
CAS Number: 333432-28-3
MDL Number: MFCD08704227
Molecular Formula: C15H15BO2
Molecular Weight: 238.09
Form: Crystal
Color: White
Formula:
C₁₅H₁₅BO₂ MW: 238.09 g/mol |
MDL Number:
MFCD08704227 CAS Number: 333432-28-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...