4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
4,4-Diethoxy-N,N-dimethylbutylamine, 4,4-Diethoxy-N,N-dimethyl-1-butanamine
D3973-25G
D3973-5G
TCD3973-25GEA
139.86
CAD
TCD3973-25G
TCD3973-5G
4-(Dimethylamino)butyraldehyde diethyl acetal ≥98.0% (by GC, titration analysis)
4-(Dimethylamino)butyraldehyde diethyl acetal
CAS Number: 1116-77-4
MDL Number: MFCD00671479
Molecular Formula: C10H23NO2
Molecular Weight: 189.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 70
Specific Gravity (20/20): 0.86
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4,4-Diethoxy-N,N-dimethylbutylamine
Formula:
C₁₀H₂₃NO₂ MW: 189.30 g/mol Boiling Pt: 95 °C Density: 0.86 (20 °C) Flash Pt: 70 °C |
MDL Number:
MFCD00671479 CAS Number: 1116-77-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...