N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation

Supplier: TCI America

Synonyms: N,N′-Di(1-naphthyl)-N,N′-diphenyl-(1,1′-biphenyl)-4,4′-diamine, N4,N4'-Di(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine

D3970-1G D3970-5G
TCD3970-1GEA 188.49 CAD
TCD3970-1G TCD3970-5G
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine ≥98.0% (by HPLC, total nitrogen), purified by sublimation
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine

CAS Number: 123847-85-8
MDL Number: MFCD03093246
Molecular Formula: C44H32N2
Molecular Weight: 588.75
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 281
Lambda max.: 339 nm (THF)

Formula: C₄₄H₃₂N₂
MW: 588.75 g/mol
Melting Pt: 281 °C
MDL Number: MFCD03093246
CAS Number: 123847-85-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bec680244defb1f438c9f78e612344e48fcc3a0&vid=ee89bd01f19ba79d674a677855c6982f9da36f20&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR