3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥95.0%
Supplier: TCI America
Synonyms:
3,6-Di(thiophen-2-yl)pyrrolo[3,4-c]pyrrole-1,4(2H,5H)-dione
D3969-1G
D3969-5G
TCD3969-1GEA
115.34
CAD
TCD3969-1G
TCD3969-5G
3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione ≥95.0%
3,6-Di(2-thienyl)-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione
CAS Number: 850583-75-4
Molecular Formula: C14H8N2O2S2
Molecular Weight: 300.35
Purity/Analysis Method: >95.0% (HPLC,N)
Form: Crystal
Formula:
C₁₄H₈N₂O₂S₂ MW: 300.35 g/mol |
MDL Number:
MFCD20257907 CAS Number: 850583-75-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...