2,5-Dibromo-1,4-dimethoxybenzene ≥98.0%

Supplier: TCI America

Synonyms: 1,4-Dibromo-2,5-dimethoxybenzene

D3966-25G D3966-5G
TCD3966-25GEA 293.88 CAD
TCD3966-25G TCD3966-5G
2,5-Dibromo-1,4-dimethoxybenzene ≥98.0%
2,5-Dibromo-1,4-dimethoxybenzene

CAS Number: 2674-34-2
MDL Number: MFCD00025694
Molecular Formula: C8H8Br2O2
Molecular Weight: 295.96
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 147


Other Product Names / Synonyms: 2,5-Dibromohydroquinone Dimethyl Ether

Formula: C₈H₈Br₂O₂
MW: 295.96
Melting Pt: 147 °C
MDL Number: MFCD00025694
CAS Number: 2674-34-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e52e7fe12312132c7af1a91e09b0762758ac7a2a&vid=22d3f61d316bfa4b769181c01b1e6dda0741613e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR