di-Sodium β-glycerophosphate pentahydrate ≥95.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

D3964-1G D3964-5G
TCD3964-1GEA 58.35 CAD
TCD3964-1G TCD3964-5G
di-Sodium β-glycerophosphate pentahydrate ≥95.0% (by titrimetric analysis) for biochemical research
di-Sodium β-glycerophosphate pentahydrate

[for Biochemical Research]
CAS Number: 13408-09-8
MDL Number: MFCD00002135
Molecular Formula: C3H9O6P
Molecular Weight: 216.04
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Alkaline Phosphatase Inhibitors (for Biochemical Research)
Other Product Names / Synonyms: beta-Glycerophosphoric Acid Disodium Salt

Formula: C₃H₇Na₂O₆P·5H₂O
MDL Number: MFCD00002135
CAS Number: 13408-09-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=caae63c2e0a5b37ea2a369c5ab1b964c8aa89553&vid=d59a1c068e20c9a763710440aa58dadd2156808c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR