1,4-Dimethyl-1,2,4-triazolium iodide ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Dimethyl-1,2,4-triazolium Iodide
D3962-5G
D3962-1G
TCD3962-5GEA
372.88
CAD
TCD3962-5G
76219-028
1,4-Dimethyl-1,2,4-triazolium iodide ≥98.0%
1,4-Dimethyl-1,2,4-triazolium iodide
CAS Number: 120317-69-3
Molecular Formula: C4H8IN3
Molecular Weight: 225.03
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 118
Formula:
C₄H₈IN₃ Melting Pt: 118 °C |
MDL Number:
MFCD00666522 CAS Number: 120317-69-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...