2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI America
D3955-1G
D3955-5G
TCD3955-1GEA
335.77
CAD
TCD3955-1G
TCD3955-5G
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid ≥97.0% (by GC, titration analysis)
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid
CAS Number: 487-66-1
Molecular Formula: C8H8O5
Molecular Weight: 184.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 99
Other Product Names / Synonyms: 2-(2-Carboxyethyl)-3-methylmaleic Anhydride
Formula:
C₈H₈O₅ Melting Pt: 99 °C |
MDL Number:
MFCD18252872 CAS Number: 487-66-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...