2,6-Dibromobenzo[1,2-b:4,5-b']dithiophene ≥98.0% (by GC)

Supplier: TCI America

D3954-1G D3954-5G
TCD3954-1GEA 388.68 CAD
TCD3954-1G TCD3954-5G
2,6-Dibromobenzo[1,2-b:4,5-b']dithiophene ≥98.0% (by GC)
2,6-Dibromobenzo[1,2-b:4,5-b']dithiophene

CAS Number: 909280-97-3
Molecular Formula: C10H4Br2S2
Molecular Weight: 348.07
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 258

Formula: C₁₀H₄Br₂S₂
MW: 348.07 g/mol
Melting Pt: 258 °C
CAS Number: 909280-97-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5642ee7d32432d0b14bcd1e4c3b2d0ea1addd14e&vid=5642ee7d32432d0b14bcd1e4c3b2d0ea1addd14e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR