3,5-Dimethyl-1H-pyrrole-2-carbaldehyde ≥98.0%

Supplier: TCI America

D3950-1G D3950-5G
TCD3950-1GEA 188.49 CAD
TCD3950-1G TCD3950-5G
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde ≥98.0%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde

CAS Number: 2199-58-8
MDL Number: MFCD00111522
Molecular Formula: C7H9NO
Molecular Weight: 123.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 91

Formula: C7H9NO
MW: 123.16 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00111522
CAS Number: 2199-58-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a42f14b6f45e79d6f917ef666e62fea95789a1f8&vid=a42f14b6f45e79d6f917ef666e62fea95789a1f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR