2,3-Diphenylmaleic anhydride ≥95.0%

Supplier: TCI America

Synonyms: 3,4-Diphenylfuran-2,5-dione

D3943-1G D3943-5G
TCD3943-1GEA 368.14 CAD
TCD3943-1G TCD3943-5G
2,3-Diphenylmaleic anhydride ≥95.0%
2,3-Diphenylmaleic anhydride

CAS Number: 4808-48-4
MDL Number: MFCD00005521
Molecular Formula: C16H10O3
Molecular Weight: 250.25
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 160


Product Categories: Analytical Science, Fluorescent Dyes
Other Product Names / Synonyms: 3,4-Diphenyl-2,5-furandione

Formula: C₁₆H₁₀O₃
Boiling Pt: 236 °C
Melting Pt: 160 °C
MDL Number: MFCD00005521
CAS Number: 4808-48-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be842a9999aadd4d31e9bc7347dc67ae595b546e&vid=be842a9999aadd4d31e9bc7347dc67ae595b546e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR