Diethyl isopropylidenemalonate ≥98.0%

Supplier: TCI America

D3942-25G D3942-5G
TCD3942-25GEA 364.98 CAD
TCD3942-25G TCD3942-5G
Diethyl isopropylidenemalonate ≥98.0%
Diethyl isopropylidenemalonate

CAS Number: 6802-75-1
MDL Number: MFCD00009147
Molecular Formula: C10H16O4
Molecular Weight: 200.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 80
Specific Gravity (20/20): 1.03
Lambda max.: 214 nm (EtOH)


Other Product Names / Synonyms: Isopropylidenemalonic Acid Diethyl Ester

Formula: C₁₀H₁₆O₄
Boiling Pt: 125 °C
Density: 1.03 (20 °C)
Flash Pt: 80 °C
MDL Number: MFCD00009147
CAS Number: 6802-75-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d286746d99f955db1cbf4abbae52b01b039719dc&vid=d286746d99f955db1cbf4abbae52b01b039719dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR