2,7-Dibromo-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
9,9-Dioctyl-2,7-dibromofluorene
D3934-25G
D3934-5G
TCD3934-25GEA
331.8
CAD
TCD3934-25G
TCD3934-5G
2,7-Dibromo-9,9-di-n-octylfluorene ≥98.0% (by HPLC)
2,7-Dibromo-9,9-di-n-octylfluorene
CAS Number: 198964-46-4
MDL Number: MFCD03427216
Molecular Formula: C29H40Br2
Molecular Weight: 548.45
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 54
Lambda max.: 280 nm (CH2Cl2)
Formula:
C₂₉H₄₀Br₂ MW: 548.45 g/mol Melting Pt: 54 °C |
MDL Number:
MFCD03427216 CAS Number: 198964-46-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...