2,7-Dibromo-9,9-dihexylfluorene ≥98.0%
Supplier: TCI America
Synonyms:
2,7-Dibromo-9,9-dihexyl-9H-fluorene
D3933-25G
D3933-5G
TCD3933-25GEA
192.76
CAD
TCD3933-25G
TCD3933-5G
2,7-Dibromo-9,9-dihexylfluorene ≥98.0%
2,7-Dibromo-9,9-dihexylfluorene
CAS Number: 189367-54-2
MDL Number: MFCD03427215
Molecular Formula: C25H32Br2
Molecular Weight: 492.34
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 68
Formula:
C₂₅H₃₂Br₂ Melting Pt: 68 °C |
MDL Number:
MFCD03427215 CAS Number: 189367-54-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...