2,7-Dibromocarbazole ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

D3932-1G D3932-5G
TCD3932-1GEA 90.06 CAD
TCD3932-1G TCD3932-5G
2,7-Dibromocarbazole ≥98.0% (by HPLC, total nitrogen)
2,7-Dibromocarbazole

CAS Number: 136630-39-2
MDL Number: MFCD09033507
Molecular Formula: C12H7Br2N
Molecular Weight: 325.00
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 232

Formula: C₁₂H₇Br₂N
Melting Pt: 232 °C
MDL Number: MFCD09033507
CAS Number: 136630-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=257e1911d373397750bee947f085456716a6b778&vid=257e1911d373397750bee947f085456716a6b778&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR