meso-1,2-Diphenylethylenediamine ≥97.0%

Supplier: TCI America

Synonyms: meso-1,2-Diphenylethane-1,2-diamine

D3930-1G D3930-5G
TCD3930-1GEA 206.66 CAD
TCD3930-1G TCD3930-5G
meso-1,2-Diphenylethylenediamine ≥97.0%
meso-1,2-Diphenylethylenediamine

CAS Number: 951-87-1
MDL Number: MFCD00274328
Molecular Formula: C14H16N2
Molecular Weight: 212.30
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 118

Formula: C₁₄H₁₆N₂
MW: 212.30 g/mol
Melting Pt: 118 °C
MDL Number: MFCD00274328
CAS Number: 951-87-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8567ef2e231cf0a89f4db3fb325c7e19a757c88&vid=e8567ef2e231cf0a89f4db3fb325c7e19a757c88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR