3,4'-Dihexyl-2,2'-bithiophene ≥96.0% (by GC)

Supplier: TCI America

D3928-1G D3928-5G
TCD3928-1GEA 387.1 CAD
TCD3928-1G TCD3928-5G
3,4'-Dihexyl-2,2'-bithiophene ≥96.0% (by GC)
3,4'-Dihexyl-2,2'-bithiophene

CAS Number: 135926-93-1
Molecular Formula: C20H30S2
Molecular Weight: 334.58
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Specific Gravity (20/20): 1.01
Lambda max.: 248 nm (THF)Storage Temperature: 0-10°C

Formula: C₂₀H₃₀S₂
Boiling Pt: 165 °C
Density: 1.01 (20 °C)
MDL Number: MFCD15072161
CAS Number: 135926-93-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58b28c2c5c8bc52afe29039030d588b25733a40c&vid=58b28c2c5c8bc52afe29039030d588b25733a40c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR