1,2-Distearoyl-sn-glycero-3-phosphocholine ≥95.0%

Supplier: TCI America

Synonyms: DSPC, 1,2-Distearoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

D3926-1G D3926-200MG
TCD3926-1GEA 486.01 CAD
TCD3926-1G TCD3926-200MG
1,2-Distearoyl-sn-glycero-3-phosphocholine ≥95.0%
1,2-Distearoyl-sn-glycero-3-phosphocholine

CAS Number: 816-94-4
MDL Number: MFCD00036905
Molecular Formula: C44H88NO8P
Molecular Weight: 790.16
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 236
Specific rotation [a]20/D: 6 deg (C=2, CHCl3)
Storage Temperature: <0°C


Product Categories: Bioscience, Lipids, Drug Delivery Systems (DDS) Research Reagents
Other Product Names / Synonyms: 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, DSPC

Formula: C₄₄H₈₈NO₈P
Melting Pt: 236 °C
MDL Number: MFCD00036905
CAS Number: 816-94-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a72fd58e982001a1734d810d7fd3357fb8893fc2&vid=a72fd58e982001a1734d810d7fd3357fb8893fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR