4,4'-Dibromo-2,2'-bipyridyl ≥97.0% (by GC, titration analysis)
Supplier: TCI America
D3919-1G
D3919-5G
D3919-25G
TCD3919-1GEA
148.84
CAD
TCD3919-1G
TCD3919-5G
TCD3919-25G
4,4'-Dibromo-2,2'-bipyridyl ≥97.0% (by GC, titration analysis)
4,4'-Dibromo-2,2'-bipyridyl
CAS Number: 18511-71-2
MDL Number: MFCD00667763
Molecular Formula: C10H6Br2N2
Molecular Weight: 313.98
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 140
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 4,4'-Dibromo-2,2'-bipyridine
Formula:
C₁₀H₆Br₂N₂ Melting Pt: 140 °C |
MDL Number:
MFCD00667763 CAS Number: 18511-71-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...