4,4'-Dinonyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3917-1G D3917-5G
TCD3917-1GEA 90.06 CAD
TCD3917-1G TCD3917-5G
4,4'-Dinonyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
4,4'-Dinonyl-2,2'-bipyridyl

CAS Number: 142646-58-0
MDL Number: MFCD00800915
Molecular Formula: C28H44N2
Molecular Weight: 408.67
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 63

Formula: C₂₈H₄₄N₂
Melting Pt: 63 °C
MDL Number: MFCD00800915
CAS Number: 142646-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=67e6307332cdbb21a542870eec7e27155758ad6d&vid=67e6307332cdbb21a542870eec7e27155758ad6d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR